Learn More
Medchemexpress LLC 3-Furanmethanol | 4412-91-3 | 98.10 | 10 G
SDP
Catalog No. 5000446586
Encompass_Preferred
3-Furanmethanol is a volatile furan compound found in malt beverages fermented with *Bifidobacterium breve*. It exhibits roasty and popcorn-like flavors and can undergo photochemical cycloaddition reactions with 2-Cyanonaphthalenes, showing high regioselectivity.
Catalog No. 50-004-46586 Supplier Medchemexpress LLC Supplier No. HYW00770810G
3-Furanmethanol is a volatile furan compound found in malt beverages fermented with *Bifidobacterium breve*. It exhibits roasty and popcorn-like flavors and can undergo photochemical cycloaddition reactions with 2-Cyanonaphthalenes, showing high regioselectivity.