Learn More
Medchemexpress LLC 3-Furanmethanol | 4412-91-3 | 98.10 | 5 G
SDP
Catalog No. 5000446585
Encompass_Preferred
3-Furanmethanol is a volatile furan compound known for its roasty, popcorn-like flavors, detectable in malt beverages fermented with *Bifidobacterium breve*. It can efficiently undergo photochemical cycloaddition reactions with 2-Cyanonaphthalenes, exhibiting high regioselectivity. This product is supplied in a liquid form with a density of 1.139 g/cm³ and appears colorless to light yellow. Solutions in DMSO require sonication and should use newly opened DMSO for optimal dissolution.
  • Identified as a flavor compound
  • Suitable for photochemical cycloaddition reactions
  • Store in pure form at -20°C for up to 3 years or 4°C for 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for 1 month
  • Soluble in DMSO and various aqueous solutions for in vitro and in vivo studies
Catalog No. 50-004-46585 Supplier Medchemexpress LLC Supplier No. HYW0077085G
3-Furanmethanol is a volatile furan compound known for its roasty, popcorn-like flavors, detectable in malt beverages fermented with *Bifidobacterium breve*. It can efficiently undergo photochemical cycloaddition reactions with 2-Cyanonaphthalenes, exhibiting high regioselectivity. This product is supplied in a liquid form with a density of 1.139 g/cm³ and appears colorless to light yellow. Solutions in DMSO require sonication and should use newly opened DMSO for optimal dissolution.
  • Identified as a flavor compound
  • Suitable for photochemical cycloaddition reactions
  • Store in pure form at -20°C for up to 3 years or 4°C for 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for 1 month
  • Soluble in DMSO and various aqueous solutions for in vitro and in vivo studies