Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Furanmethanol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4412-91-3
C5H6O2
98.1
MFCD00005352
STJIISDMSMJQQK-UHFFFAOYSA-N
3-furanmethanol,furan-3-methanol,3-furylmethanol,3-hydroxymethyl furan,3-furfuryl alcohol,3-hydroxymethylfuran,3-furyl alcohol,furan-3-yl-methanol,furan-3-yl methanol,3-hydroxymethyl-furan
20449
furan-3-ylmethanol
C1=COC=C1CO
4412-91-3
C5H6O2
98.1
MFCD00005352
STJIISDMSMJQQK-UHFFFAOYSA-N
3-furanmethanol,furan-3-methanol,3-furylmethanol,3-hydroxymethyl furan,3-furfuryl alcohol,3-hydroxymethylfuran,3-furyl alcohol,furan-3-yl-methanol,furan-3-yl methanol,3-hydroxymethyl-furan
20449
furan-3-ylmethanol
C1=COC=C1CO
Specific Gravity 1.139
Boiling Point 79.0°C to 80.0°C (17.0mmHg)
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4835 to 1.4855
Quantity 5g
Density 1.1390g/mL
Flash Point 38°C
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Beilstein 17, III, 1266
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 98.1
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Furanmethanol
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cauti

GHS Signal Word: Warning

EINECSNumber : 224-570-7