Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Furo[3,2-b]pyridin-6-yl)prop-2-yn-1-ol

Catalog No. ADE0011531G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0011531G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0011531G Supplier Sigma Aldrich Supplier No. ADE0011531G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Indole-3-propionic acid can be obtained from tryptophan by deamination reaction.reactant for preparation of:
  • Fluorescent analogues of strigolactones
  • Anti-tumor agents
  • Melanocortin receptors ligands
  • Immunosuppressive agents
  • Iinhibitors of hepatitis c virus
  • Histamine h4 receptor agonists
  • Nr2b/nmda receptor antagonists
  • Cb1 antagonist for the treatment of obesity
  • Antibacterial agents
  • Inhibitor of tgf-α receptor bindingindole-3-propionic acid may be used in the synthesis of oxindole-3-propionic acid via reaction with n-bromosuccinimide in acetic acid followed by treatment with h2/pd catalyst.studied as an adjunct to improve perfusion after liver transplant

Specifications

Molecular Formula C10H7NO2
Linear Formula C10H7NO2
MDL Number MFCD15530315
Quantity 25 g
Molecular Weight (g/mol) 173.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.