Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 3-Furoic acid, 488-93-7, MFCD00005350, 25g

Catalog No. 5001379060
Encompass_Preferred

Molecular Formula C5H4O3, Purity 97%, Molecular Weight 112.09, Melting Point 122-124°

Catalog No. 50-013-79060 Supplier Matrix Scientific Supplier No. 03746125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C5H4O3, Purity 97%, Molecular Weight 112.09, Melting Point 122-124°

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.