Learn More
Apexbio Technology LLC 3-FUROIC-ACID-500MG
SDP
Catalog No. 5000569964
Encompass_Preferred
3-Furoic acid (CAS No 488-93-7) is a compound used extensively in biomedical research due to its versatile applications Derived from furan it is often explored for its role in modulating biochemical pathways involved in oxidative stress and inflammation Researchers have observed its activity within the nanomolar to low micromolar range in various in vitro studies Common applications of 3-Furoic acid include drug screening where it is used to explore its effects on different cell models and in animal studies to investigate its potential therapeutic benefits Its mechanism of action is believed to involve interference with specific targets such as enzymes and signaling pathways making it a subject of interest for developing anti-inflammatory and antioxidant therapies Experimental concentrations typically depend on the specific research design allowing flexibility in study protocols
Catalog No. 50-005-69964 Supplier Apexbio Technology LLC Supplier No. C7174500
3-Furoic acid (CAS No 488-93-7) is a compound used extensively in biomedical research due to its versatile applications Derived from furan it is often explored for its role in modulating biochemical pathways involved in oxidative stress and inflammation Researchers have observed its activity within the nanomolar to low micromolar range in various in vitro studies Common applications of 3-Furoic acid include drug screening where it is used to explore its effects on different cell models and in animal studies to investigate its potential therapeutic benefits Its mechanism of action is believed to involve interference with specific targets such as enzymes and signaling pathways making it a subject of interest for developing anti-inflammatory and antioxidant therapies Experimental concentrations typically depend on the specific research design allowing flexibility in study protocols