Learn More
Sigma Organic Chemistry 3-Furoyl chloride | 5G | 26214-65-3 | MFCD02681970
Supplier: Sigma Organic Chemistry 7156895G

3-Furoyl chloride , 5G
About this item:
CAS #: 26214-65-3
MDL #: MFCD02681970
Purity: ≥97
Molecular Weight: 130.53
UNSPSC Code: 12352100
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.