Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Furylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F04385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0438-5G 5 g
F04381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0438-5G Supplier TCI America Supplier No. F04385G
Add to Cart
Add to Cart

Specifications

CAS 55552-70-0
Color White-Yellow
Molecular Formula C4H5BO3
Quantity 5 g
MDL Number MFCD01319007
Synonym furan-3-boronic acid, 3-furanboronic acid, 3-furylboronic acid, furan-3-yl boronic acid, 3-furanylboronic acid
InChI Key CYEFKCRAAGLNHW-UHFFFAOYSA-N
SMILES B(C1=COC=C1)(O)O
IUPAC Name furan-3-ylboronic acid
Molecular Weight (g/mol) 111.891
PubChem CID 2734358
Formula Weight 111.89
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.