Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab 3% Glutaraldehyde, 5 mL Prefills
SDP

Catalog No. 22038183
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-038-183 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-038-183 Supplier StatLab Supplier No. FXGLU35CS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Leading fixative for electron microscopy.

  • A strong chemical fixative to preserve fine cellular structures and ultrastructural details for electron microscopy
  • 5 mL prefills
  • 50/Cs.

Specifications

Packaging Type Prefills
For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 5 mL
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.