Learn More
StatLab 3% Glutaraldehyde Cacodylate Solution

Description
- 3% Glutaraldehyde in cacodylate buffer (pH 7.3)
- Commonly used as a primary fixative for electron microscopy
- An effective fixative that forms cross-links between proteins, stabilizing the structure of cells and tissues which allows them to survive the process of preparation and staining without degradation or structural changes
- 5 mL prefilled containers, 50/Pk.
Specifications
Specifications
| For Use With (Equipment) | All Tissue Processors |
| Grade | Chemical Solution |
| Product Type | Fixative |
| Solution Type | Fixation Buffer |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.