Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab 3% Glutaraldehyde Fixative
SDP

Catalog No. 23640178
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
23-640-178 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-178 Supplier StatLab Supplier No. FXGLU3PT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

3% Glutaraldehyde is a leading fixative for Electron Microscopy and the preservation of ultra structures

  • Glutaraldehyde 3% solution forms cross-links between proteins, preserving biological tissue specimens and allowing them to survive the process of preparation and staining without degradation or structural changes

Specifications

Grade Histological
Product Type Fixative
Quantity 1 p.
Solution Type Fixation Buffer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.