Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Glycidyloxypropyl)Triethoxysilane

Catalog No. 5005950ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-059-50ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 50-059-50ML Supplier Sigma Aldrich Supplier No. 5005950ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Glycidyloxypropyl)triethoxysilane (GPTES) can be used as a reagent in the synthesis of: Polymer nanocomposite membranes for direct methanol fuel cells (DMFCs) Azido terminated poly(ethylene glycol) silane that can be self-assembled on a metal-oxide surface to facilitate the orthogonal biofunctionalization Epoxy functionalized silsesquioxane nanoparticles (SQ-NPs). Epoxy-functionalized mesoporous cellular foams (G-MCFs) as the support for the immobilization of penicillin G acylase (PGA). GPTES can also be used as a precursor to prepare water-repellent, self-cleaning coatings.

Specifications

CAS 18543-68-5
Molecular Formula C10H16MgN2O8 -+ 4H2O
Refractive Index n20/D 1.427
Linear Formula C10H16MgN2O8 -+ 4H2O
MDL Number MFCD00150929
Quantity 50 mL
Synonym GPTES; 3-(2,3-Epoxypropyloxy)propyltriethoxysilane
Molecular Weight (g/mol) 388.61 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.