Learn More
U.S. Pharmacopeia Sodium Fluoride, 7681-49-4, MFCD00003524, 1g
Catalog No. NC9093370
Encompass
Molecular formula NaF, Molecular weight 41.99, Melting Point 1819.4 °F (993 °C), Boiling Point 3099.2 °F (1704 °C)
Catalog No. NC9093370 Supplier U.S. Pharmacopeia Supplier No. 1614002
Molecular formula NaF, Molecular weight 41.99, Melting Point 1819.4 °F (993 °C), Boiling Point 3099.2 °F (1704 °C)