Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Heptyn-1-ol 97.0+%, TCI America™
SDP

Catalog No. H04575ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04575ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04575ML Supplier TCI America Supplier No. H04575ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14916-79-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00039551
InChI Key PSWHODJVUOXHKA-UHFFFAOYSA-N
Synonym 3-heptyn-1-ol, acmc-209d1z, ho ch2 2c=c ch2 2ch3
PubChem CID 84693
IUPAC Name hept-3-yn-1-ol
SMILES CCCC#CCCO

Specifications

Color Yellow
Boiling Point 111°C
Quantity 5 mL
Formula Weight 112.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Heptyn-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.