Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Hexylthiophene-2-Boronic Acid Pinacol Ester

Catalog No. 6974005G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-740-05G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-740-05G Supplier Sigma Aldrich Supplier No. 6974005G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used forSuzuki-Miyaura cross-coupling reactions p-type/n-type switching of ambipolar bithiazole-benzothiadiazole-based polymers in solar cells Hierarchical self-assembly of semiconductor functionalized peptide a-helixes and optoelectronic properties Reagent used in Preparation ofPhotovoltaic materials, polymers, and thiophene-based compounds with photophysical, electrochemical, and fluorescent propertiesPolymer solar cells for Low band gap poly(1,4-arylene-2,5-thienylene)s with benzothiadiazole units Dithienothiophene-based dyes for dye-sensitized solar cells

Specifications

CAS 64417-12-5
Density 0.811 g/mL at 25°C
Molecular Formula C12H20
Refractive Index n20/D 1.490-1.499
Linear Formula C12H20
MDL Number MFCD00145386
Quantity 1 g
Synonym 2-(3-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 3-Hexyl-2-thienylboronic acid
Molecular Weight (g/mol) 164.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.