Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hexylthiophene 99+%, Thermo Scientific™

Catalog No. AC296120100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC296120100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC296120100 Supplier Thermo Scientific Chemicals Supplier No. 296120100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1693-86-3
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.298
InChI Key JEDHEMYZURJGRQ-UHFFFAOYSA-N
Synonym 3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
PubChem CID 566849
IUPAC Name 3-hexylthiophene
SMILES CCCCCCC1=CSC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow or Red
Density 0.936
Infrared Spectrum Authentic
Refractive Index 1.4950 to 1.4970 (20°C, 589nm)
Quantity 10 g
Formula Weight 168.3
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material 3-Hexylthiophene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.