Learn More
Grainger 3 Hole - Center Tap Base Plate Double
Catalog No. 502766481
Encompass_Preferred
The Tripp Lite 16U351 is a 19-inch open frame rack enclosure cabinet with a depth of 35 inches. It has an adjustable mounting depth from 22 to 35 inches and comes with mounting rails, leveling feet and a vented top panel. The steel enclosure has a weight capacity of 1000 lbs and is compatible with EIA-310-D mounting rails. The cabinet comes unassembled in a flat pack box to reduce shipping costs. Overall, the 16U351 provides an open and adaptable rack enclosure solution for mounting up to 16U of 19-inch rackmount equipment.
Catalog No. 50-276-6481 Supplier Grainger Supplier No. 16U351
The Tripp Lite 16U351 is a 19-inch open frame rack enclosure cabinet with a depth of 35 inches. It has an adjustable mounting depth from 22 to 35 inches and comes with mounting rails, leveling feet and a vented top panel. The steel enclosure has a weight capacity of 1000 lbs and is compatible with EIA-310-D mounting rails. The cabinet comes unassembled in a flat pack box to reduce shipping costs. Overall, the 16U351 provides an open and adaptable rack enclosure solution for mounting up to 16U of 19-inch rackmount equipment.