Learn More
METTLER TOLEDO 3P INDUSTRIAL 3-HOLE STOPPERS FOR TITRATORS
Catalog No. NC9085892
Encompass_Preferred
Stoppers Set for Three-Hole Adapter, Comprising: 1 ST10 Stopper for DL35 Karl Fischer Titrator
Catalog No. NC9085892 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 25883
Stoppers Set for Three-Hole Adapter, Comprising: 1 ST10 Stopper for DL35 Karl Fischer Titrator