Learn More
Medchemexpress LLC 3-hydrazinoquinoxaline-2-thiol | 13080-21-2 | 98.0% | 5 MG
SDP
Catalog No. 5000234565
Encompass_Preferred
HPi1 is a potent, selective, orally active antimicrobial research compound targeting Helicobacter pylori. It demonstrates an IC50 of 0.24 μM and an MIC of 0.08-0.16 μg/mL, and is supplied as a solid research-grade material for laboratory use.
  • Potent activity vs Helicobacter pylori (IC50 = 0.24 μM).
  • Low minimum inhibitory concentration (MIC = 0.08-0.16 μg/mL).
  • High purity (98.0%).
  • Solid form, light yellow to yellow.
  • Molecular formula C8H8N4S; molecular weight 192.24 g/mol.
  • Available in small research pack sizes, including 5 mg.
Catalog No. 50-002-34565 Supplier Medchemexpress LLC Supplier No. HY1205365MG
HPi1 is a potent, selective, orally active antimicrobial research compound targeting Helicobacter pylori. It demonstrates an IC50 of 0.24 μM and an MIC of 0.08-0.16 μg/mL, and is supplied as a solid research-grade material for laboratory use.
  • Potent activity vs Helicobacter pylori (IC50 = 0.24 μM).
  • Low minimum inhibitory concentration (MIC = 0.08-0.16 μg/mL).
  • High purity (98.0%).
  • Solid form, light yellow to yellow.
  • Molecular formula C8H8N4S; molecular weight 192.24 g/mol.
  • Available in small research pack sizes, including 5 mg.