Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester, Thermo Scientific™

Catalog No. aah2596322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2596322 100 g
AAH2596336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2596322 Supplier Thermo Scientific Chemicals Supplier No. H2596322

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester is a reactant in the Tischenko Reaction.

Solubility
Soluble in methanol(almost transparency).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-20-4
Molecular Formula C10H20O4
Molecular Weight (g/mol) 204.266
MDL Number MFCD00059597
InChI Key SZCWBURCISJFEZ-UHFFFAOYSA-N
Synonym hydroxypivalic acid neopentyl glycol ester, esterdiol 204, 3-hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropionate, hydroxyneopentyl hydroxypivalate, unii-cg0a37gahj, propanoic acid, 3-hydroxy-2,2-dimethyl-, 3-hydroxy-2,2-dimethylpropyl ester, neopentyl glycol monohydroxypivalate, 3-hydroxypivaloyloxy-2,2-dimethylpropanol, 3-hydroxy-2,2-dimethylpropyl hydroxypivalate, neopental glycol monohydroxypivalate
PubChem CID 14218
IUPAC Name (3-hydroxy-2,2-dimethylpropyl) 3-hydroxy-2,2-dimethylpropanoate
SMILES CC(C)(CO)COC(=O)C(C)(C)CO

Specifications

Melting Point 51°C to 54°C
Density 1.01
Boiling Point 292°C
Flash Point 161°C (322°F)
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in methanol(almost transparency).
Formula Weight 204.27
Chemical Name or Material 3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
EINECSNumber 214-222-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.