Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2-iodopyridine 98.0+%, TCI America™
SDP

Catalog No. H13961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H13961G Supplier TCI America Supplier No. H13961G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity 3-Hydroxy-2-iodopyridine laboratory reagent with 98.0% GC purity.

Chemical Identifiers

CAS 40263-57-8
Molecular Formula C5H4INO
Molecular Weight (g/mol) 220.997
MDL Number MFCD00023421
InChI Key HJBGMPCMSWJZNH-UHFFFAOYSA-N
Synonym 2-iodo-3-hydroxypyridine, 2-iodo-3-pyridinol, 3-hydroxy-2-iodopyridine, 3-pyridinol, 2-iodo, 2-iodo-pyridin-3-ol, 2-iodpyridin-3-ol, 2-iodopyridine-3-ol, acmc-209jcf, 2-iodo-3-hydroxypyridin, 2-iodo-3-hydroxy-pyridine
PubChem CID 97179
IUPAC Name 2-iodopyridin-3-ol
SMILES C1=CC(=C(N=C1)I)O

Specifications

Melting Point 189°C
Color Yellow
Quantity 1 g
Formula Weight 221.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.