Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Hydroxy-2-naphthoic acid

Catalog No. H46007100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H46007100G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H46007100G Supplier Sigma Aldrich Supplier No. H46007100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant for:
  • Nickel-catalyzed alkyl-alkyl suzuki coupling reactions with boron reagents
  • Preparation of a selective small-molecule melanocortin-4 receptor agonist with efficacy in a pilot study of sexual dysfunction in humans
  • Preparation of aliphatic hydrocarbons via nickel-catalyzed suzuki cross-coupling with alkylboranes
  • Preparation of anthranilic acids as antibacterial agents with human serum albumin binding affinity
  • Preparation of antiatherogenic antioxidant di-tert-butyldihydrobenzofuranols via grignard reactions with di-tert-butyl(hydroxy)benzaldehyde derivatives
  • Synthesis of gephyrotoxin via the schmidt reaction

Specifications

CAS 92-70-6
Melting Point 218°C to 221°C (lit.)
Molecular Formula C11H8O3
Linear Formula HOC10H6CO2H
MDL Number MFCD00004103
Quantity 500 mg
Synonym beta-Hydroxynaphthoic acid
Molecular Weight (g/mol) 188.18
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.