Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2-naphthoic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
Packaging:
Glass bottle
Plastic bottle
92-70-6
C11H8O3
188.18
MFCD00004103
ALKYHXVLJMQRLQ-UHFFFAOYSA-N
3-hydroxy-2-naphthoic acid,2-hydroxy-3-naphthoic acid,bon acid,bona,developer bon,2-naphthol-3-carboxylic acid,2-naphthalenecarboxylic acid, 3-hydroxy,b.o.n. acid,c.i. developer 20,miketazol developer ons
7104
CHEBI:80383
3-hydroxynaphthalene-2-carboxylic acid
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)O
92-70-6
C11H8O3
188.18
MFCD00004103
ALKYHXVLJMQRLQ-UHFFFAOYSA-N
3-hydroxy-2-naphthoic acid,2-hydroxy-3-naphthoic acid,bon acid,bona,developer bon,2-naphthol-3-carboxylic acid,2-naphthalenecarboxylic acid, 3-hydroxy,b.o.n. acid,c.i. developer 20,miketazol developer ons
7104
CHEBI:80383
3-hydroxynaphthalene-2-carboxylic acid
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)O
Quantity 100g
Melting Point 217°C to 223°C
Flash Point >150°C
Packaging Plastic bottle
Infrared Spectrum Authentic
Color Beige-Brown to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 10, 333
Merck Index 15, 4874
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alkalies,freely soluble in alcohol,ether and benzene
Formula Weight 188.18
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Hydroxy-2-naphthoic acid, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-180-8

RTECSNumber : QL1755000

TSCA : TSCA