Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-3-methyl-2-butanone Oxime 98.0+%, TCI America™
SDP

Catalog No. H047225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H047225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H047225G Supplier TCI America Supplier No. H047225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7431-25-6
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00059656
InChI Key QKLLBCGVADVPKD-XQRVVYSFSA-N
Synonym 3-hydroxy-3-methyl-2-butanone oxime, 3e-3-hydroxyimino-2-methylbutan-2-ol, qkllbcgvadvpkd-xqrvvysfsa, 3-hydroxy-3-methyl-butan-2-one oxime, 3z-3-hydroxyimino-2-methylbutan-2-ol, 3e-3-hydroxyimino-2-methyl-butan-2-ol, 2z-3-hydroxy-3-methyl-2-butanone oxime #
PubChem CID 5385758
IUPAC Name (3Z)-3-hydroxyimino-2-methylbutan-2-ol
SMILES CC(=NO)C(C)(C)O

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 25 g
Formula Weight 117.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone Oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.