Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Hydroxy-3-methylbutyronitrile

Catalog No. 5562450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-624-50ML 50 mL
55-624-10ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-624-50ML Supplier Sigma Aldrich Supplier No. 5562450ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-hydroxy-3-methylbutyronitrile is a α-hydroxynitrile
  • It undergoes thermal degradation in gas phase via a six-membered cyclic transition state
  • 3-hydroxy-3-methylbutyronitrile can be synthesized from 2-hydroxy-2-methyl-1-bromopropane.3-hydroxy-3-methylbutyronitrile may be used to synthesize 1,1-dimethylcyanoethyl bromoacetate

Specifications

CAS 13635-04-6
Density 0.959 g/mL (at 20°C (literature))
Boiling Point 114°C to 116°C (30 mmHg)
Molecular Formula C5H9NO
Linear Formula HOC(CH3)2 CH2CN
Refractive Index n20/D 1.430
MDL Number MFCD00043123
Quantity 50 mL
Synonym beta-Hydroxyisovaleronitrile
Molecular Weight (g/mol) 99.13
Percent Purity ≥97.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.