Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-3-phenylpropionic Acid 98.0+%, TCI America™
SDP

Catalog No. H13945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13945G 5 g
H13941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13945G Supplier TCI America Supplier No. H13945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3480-87-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00137907
InChI Key AYOLELPCNDVZKZ-UHFFFAOYSA-N
PubChem CID 92959
ChEBI CHEBI:19929
IUPAC Name 3-hydroxy-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)O

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-3-phenylpropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.