Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
3-Hydroxy-4-methoxybenzaldehyde
Spécifications
Spécifications
| CAS | 621-59-0 |
| Melting Point | 113°C to 115°C (lit.) |
| Boiling Point | 179°C (15 mmHg) |
| Molecular Formula | C8H8O3 |
| Linear Formula | HOC6H3(OCH3)CHO |
| Quantity | 25 g |
| MDL Number | MFCD00003369 |
| Synonym | 3-Hydroxyanisaldehyde; Isovanillin |
| Molecular Weight (g/mol) | 152.15 |
| Percent Purity | 99% |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'