Learn More
Medchemexpress LLC HY-N0761 100mg Medchemexpress, 3-Hydroxy-4-methoxycinnamic acid CAS:537-73-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0761100MG
Encompass_Preferred
Medchemexpress, HY-N0761 100mg 3-Hydroxy-4-methoxycinnamic acid CAS:537-73-5 3-Hydroxy-4-methoxycinnamic acid (Isoferulic acid) is a cinnamic acid derivative that has antidiabetic activity. 3-Hydroxy-4-methoxycinnamic acid binds to and activates α1-adrenergic receptors (IC 50 =1.4 µM) to enhance secretion of β-endorphin (EC 50 =52.2 nM) and increase glucose use. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3703