Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Hydroxy-4-methoxyphenylacetic acid

Catalog No. 7163911G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-639-11G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-639-11G Supplier Sigma Aldrich Supplier No. 7163911G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,7-octadiene can serve as a crosslinker and source of ethylene for a variant of mori's conditions in ceym-related reactions.1,7-octadiene has been used in a study to assess the structure and reaction rate in olefin ring-closing metathesis of a series of simple dienes
  • It has also been used in a study to investigate micropatterned surfaces prepared by plasma polymerization

Specifications

CAS 1131-94-8
Melting Point 127°C to 132°C
Molecular Formula C9H10O4
Linear Formula C9H10O4
MDL Number MFCD00016829
Quantity 100 g
Molecular Weight (g/mol) 182.17
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.