Learn More
Sigma Aldrich 3-Hydroxy-4-methoxyphenylboronic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1,2,3-triazines have been shown to be reactive substrates in inverse electron demand diels-alder strategies
- Recent examples by the-ßboger research group-ßhave utilized this reactive motif in the construction of highly functionalized n-containing heterocycles.total syntheses of (-)-pyrimidoblamic acid and p-3a
Specifications
Specifications
| Quantity | 100 mg |
| Molecular Formula | C7H9BO4 |
| Linear Formula | C7H9BO4 |
| MDL Number | MFCD12025997 |
| Molecular Weight (g/mol) | 167.95 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.