Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-methoxyphenylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AC445060010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445060010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445060010 Supplier Thermo Scientific Chemicals Supplier No. 445060010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 269410-23-3
Molecular Formula C13H19BO4
Molecular Weight (g/mol) 250.10
MDL Number MFCD09027077
InChI Key WINSAMQIAMEZIK-UHFFFAOYSA-N
Synonym 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxy-4-methoxyphenylboronic acid, pinacol ester, 2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxy-4-methoxyphenylboronic acid pinacol ester, phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, amtb953, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenol
PubChem CID 17750251
IUPAC Name 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES COC1=CC=C(C=C1O)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 101.0°C to 102.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 250.1
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-4-methoxyphenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.