Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-nitrobenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. H13345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13345G 5 g
H13341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13345G Supplier TCI America Supplier No. H13345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 704-13-2
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007109
InChI Key AUBBVPIQUDFRQI-UHFFFAOYSA-N
Synonym benzaldehyde, 3-hydroxy-4-nitro, 3-hydroxy-4-nitro-benzaldehyde, 4-nitro-3-hydroxybenzaldehyde, 3-formyl-6-nitrophenol, acmc-1bm06, ksc495a6r, 3-hydroxy-4-nitro benzaldehyde, 3-hydroxy-4-nitrobenzaldehyde
PubChem CID 69712
IUPAC Name 3-hydroxy-4-nitrobenzaldehyde
SMILES OC1=CC(C=O)=CC=C1[N+]([O-])=O

Specifications

Melting Point 131°C
Quantity 5 g
Formula Weight 167.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4-nitrobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.