Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Hydroxy-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid

Catalog No. CDS023983 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS023983 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS023983 Supplier Sigma Aldrich Supplier No. CDS02398325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant for:
  • Oxidative cyclization with chlorine to give chloro- and acetoxybenziodoxaboroles and a self-assembled tetrameric macrocyclic structure
  • Preparation of allylic alcohols or enone derivatives using boronic acid as catalystcatalyst for:
  • Preparation of acylsulfoximines via n-acylation of sulfoximines with aliphatic acids
  • Direct amide bond formation
  • Regioselective preparation of substituted triazole derivatives via dipolar cycloaddition of unsaturated carboxylic acids with azides

Specifications

Molecular Formula C11H12O3
Linear Formula C11H12O3
MDL Number MFCD13368205
Quantity 500 mg
Molecular Weight (g/mol) 192.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.