Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-6-methoxyflavone 98.0+%, TCI America™
SDP

Catalog No. H15221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15221G 1 g
H15225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15221G Supplier TCI America Supplier No. H15221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93176-00-2
Molecular Formula C16H12O4
Molecular Weight (g/mol) 268.268
MDL Number MFCD00017639
InChI Key OGURJSOPVFCIOO-UHFFFAOYSA-N
Synonym 3-hydroxy-6-methoxyflavone, 6-methoxyflavonol, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-one, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-on, 4h-1-benzopyran-4-one, 3-hydroxy-6-methoxy-2-phenyl, 3-hydroxy-6-methoxy-2-phenyl-4h-1-benzopyran-4-one
PubChem CID 688676
IUPAC Name 3-hydroxy-6-methoxy-2-phenylchromen-4-one
SMILES COC1=CC2=C(C=C1)OC(=C(C2=O)O)C3=CC=CC=C3

Specifications

Melting Point 208°C
Color White-Yellow
Quantity 1 g
Formula Weight 268.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-6-methoxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.