Learn More
Medchemexpress LLC 3α-hydroxy-7-oxo-5β-cholanic acid | 4651-67-6 | MFCD00271393 | >98.0% | 390.6 g/mol | C24H38O4 | 100 MG
SDP
Catalog No. 5000301295
Encompass_Preferred
7-Ketolithocholic acid is a bile acid analytical standard (3α-hydroxy-7-oxo-5β-cholanic acid) used in research and analytical applications to study bile acid metabolism and related pathways. The material is provided at high purity for reliable assay performance in chromatographic and spectrometric methods.
  • Analytical standard for bile acid research.
  • High purity suitable for chromatography and mass spectrometry.
  • Molecular formula C24H38O4 and molecular weight 390.6 g/mol.
  • Available in small pack sizes for analytical workflows.
  • Supports studies of bile acid metabolism and liver function.
Catalog No. 50-003-01295 Supplier Medchemexpress LLC Supplier No. HYW018512R100MG
7-Ketolithocholic acid is a bile acid analytical standard (3α-hydroxy-7-oxo-5β-cholanic acid) used in research and analytical applications to study bile acid metabolism and related pathways. The material is provided at high purity for reliable assay performance in chromatographic and spectrometric methods.
  • Analytical standard for bile acid research.
  • High purity suitable for chromatography and mass spectrometry.
  • Molecular formula C24H38O4 and molecular weight 390.6 g/mol.
  • Available in small pack sizes for analytical workflows.
  • Supports studies of bile acid metabolism and liver function.