Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3α-hydroxy-7-oxo-5β-cholanic acid | 4651-67-6 | MFCD00271393 | 99.9% | 390.56 g/mol | C24H38O4 | 1 G
SDP

Catalog No. 5000301869
Encompass_Preferred

7-Ketolithocholic acid (3α-hydroxy-7-oxo-5β-cholanic acid) is a bile acid derivative used as a research reagent and analytical standard. It is reported to be absorbed and to suppress endogenous bile acid production and biliary cholesterol secretion. The compound is provided as a high-purity solid and as ready-to-use DMSO solutions for laboratory experiments.

  • High purity: 99.93%.
  • Chemical formula: C24H38O4.
  • Molecular weight: 390.56 g/mol.
  • Available as solid and as 10 mM solutions in DMSO.
  • Suitable as a reference standard and research reagent.
  • CAS number: 4651-67-6.

Catalog No. 50-003-01869 Supplier Medchemexpress LLC Supplier No. HYW0185121G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Ketolithocholic acid (3α-hydroxy-7-oxo-5β-cholanic acid) is a bile acid derivative used as a research reagent and analytical standard. It is reported to be absorbed and to suppress endogenous bile acid production and biliary cholesterol secretion. The compound is provided as a high-purity solid and as ready-to-use DMSO solutions for laboratory experiments.

  • High purity: 99.93%.
  • Chemical formula: C24H38O4.
  • Molecular weight: 390.56 g/mol.
  • Available as solid and as 10 mM solutions in DMSO.
  • Suitable as a reference standard and research reagent.
  • CAS number: 4651-67-6.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.