Learn More
Medchemexpress LLC 3-hydroxy agomelatine | 166526-99-4 | MFCD27996054 | 99.1% | 259.31 g/mol | C15H17NO3 | 5 MG
SDP
Catalog No. 5000262494
Encompass_Preferred
3-Hydroxy agomelatine is a research-grade metabolite of agomelatine used in receptor pharmacology and metabolic studies. It acts as a 5-HT2C receptor antagonist and is supplied as an off-white to light-brown solid suitable for analytical and in vitro assays. Common applications include affinity profiling, metabolite identification, and impurity characterization.
  • Acts as a 5-HT2C receptor antagonist (IC50 ≈ 3.2 μM; Ki ≈ 1.8 μM).
  • Supplied as an off-white to light-brown solid for ease of handling.
  • High reported purity (about 99.1%) suitable for research applications.
  • Useful as a reference standard for metabolic and impurity studies.
  • Available in small-mass formats and DMSO solutions for assay setup.
Catalog No. 50-002-62494 Supplier Medchemexpress LLC Supplier No. HY1331115MG
3-Hydroxy agomelatine is a research-grade metabolite of agomelatine used in receptor pharmacology and metabolic studies. It acts as a 5-HT2C receptor antagonist and is supplied as an off-white to light-brown solid suitable for analytical and in vitro assays. Common applications include affinity profiling, metabolite identification, and impurity characterization.
  • Acts as a 5-HT2C receptor antagonist (IC50 ≈ 3.2 μM; Ki ≈ 1.8 μM).
  • Supplied as an off-white to light-brown solid for ease of handling.
  • High reported purity (about 99.1%) suitable for research applications.
  • Useful as a reference standard for metabolic and impurity studies.
  • Available in small-mass formats and DMSO solutions for assay setup.