Learn More
Cayman Chemical 3-hydroxy-DL-Kynurenine, Purity: ≥98%, CAS: 484-78-6, 25 mg
SDP
Catalog No. 502941213
Encompass_Preferred
An active metabolite of tryptophan; inhibits yeast and rat liver aldehyde dehydrogenase by 97 and 69%, respectively, at 100 μM; active against MRSA, S. epidermidis, VRE, E. coli, MDRP, and C. albicans (IC50s = 31.2, 39.2, 57.6, 24, 25.6, and 137.6 μg/ml, respectively); increases intracellular NAD+ levels and extracellular LDH activity in human neurons and astrocytes
Catalog No. 50-294-1213 Supplier Cayman Chemical Supplier No. 2777825 MG
An active metabolite of tryptophan; inhibits yeast and rat liver aldehyde dehydrogenase by 97 and 69%, respectively, at 100 μM; active against MRSA, S. epidermidis, VRE, E. coli, MDRP, and C. albicans (IC50s = 31.2, 39.2, 57.6, 24, 25.6, and 137.6 μg/ml, respectively); increases intracellular NAD+ levels and extracellular LDH activity in human neurons and astrocytes