Learn More
Cayman Chemical 3-hydroxy Lauric Acid, Purity: ≥98%, CAS: 1883-13-2, 10 mg
SDP
Catalog No. 502936293
Encompass_Preferred
A hydroxy fatty acid; found in bacteria as well as Pseudosuberites and D. calyx sea sponges; antifungal activity against a panel of seven fungi (MICs = 10-50 μg/ml); partial agonist of GPR84 receptors in vitro (EC50 = 5.24 μM)
Catalog No. 50-293-6293 Supplier Cayman Chemical Supplier No. 2464210 MG
A hydroxy fatty acid; found in bacteria as well as Pseudosuberites and D. calyx sea sponges; antifungal activity against a panel of seven fungi (MICs = 10-50 μg/ml); partial agonist of GPR84 receptors in vitro (EC50 = 5.24 μM)