Learn More
Cayman Chemical 3'-hydroxy Lidocaine, Purity: ≥98%, CAS: 34604-55-2, 10 mg
SDP
Catalog No. 502964535
Encompass_Preferred
A metabolite of lidocaine; formed by CYP1A2, CYP2D6, and CYP3A4
Catalog No. 50-296-4535 Supplier Cayman Chemical Supplier No. 2289910 MG
A metabolite of lidocaine; formed by CYP1A2, CYP2D6, and CYP3A4