Learn More
Cayman Chemical 3-hydroxy LIgnocerIc AcId 1mg
SDP
Catalog No. 502995613
Encompass_Preferred
A long-chain saturated fatty acid; found in minor amounts in Gram-negative bacterial isolates; intermediate in fatty acid chain elongation, in the form of an acyl-CoA metabolite
Catalog No. 50-299-5613 Supplier Cayman Chemical Supplier No. 226671 MG
A long-chain saturated fatty acid; found in minor amounts in Gram-negative bacterial isolates; intermediate in fatty acid chain elongation, in the form of an acyl-CoA metabolite