Learn More
Cayman Chemical 3-hydroxy Palmitic Acid, Purity: ≥95%, CAS: 2398-34-7, 100 mg
SDP
Catalog No. 502985005
Encompass_Preferred
Found in the lipid A part of LPS and useful as chemical marker of endotoxins; accumulates during long-chain 3-hydroxy-acyl-CoA dehydrogenase and mitochondrial trifunctional protein deficiencies, inducing oxidative stress and leading to mitochondrial bioenergetics deregulation and eventual multi-organ dysfunction
Catalog No. 50-298-5005 Supplier Cayman Chemical Supplier No. 19934100 MG
Found in the lipid A part of LPS and useful as chemical marker of endotoxins; accumulates during long-chain 3-hydroxy-acyl-CoA dehydrogenase and mitochondrial trifunctional protein deficiencies, inducing oxidative stress and leading to mitochondrial bioenergetics deregulation and eventual multi-organ dysfunction