Learn More
Cayman Chemical 3-hydroxy PalmItIc AcId 250mg
SDP
Catalog No. 502982859
Encompass_Preferred
Found in the lipid A part of LPS and useful as chemical marker of endotoxins; accumulates during long-chain 3-hydroxy-acyl-CoA dehydrogenase and mitochondrial trifunctional protein deficiencies, inducing oxidative stress and leading to mitochondrial bioenergetics deregulation and eventual multi-organ dysfunction
Catalog No. 50-298-2859 Supplier Cayman Chemical Supplier No. 19934250 MG
Found in the lipid A part of LPS and useful as chemical marker of endotoxins; accumulates during long-chain 3-hydroxy-acyl-CoA dehydrogenase and mitochondrial trifunctional protein deficiencies, inducing oxidative stress and leading to mitochondrial bioenergetics deregulation and eventual multi-organ dysfunction