Learn More
Medchemexpress LLC 3α-Hydroxy pravastatin sodium | 81093-43-8 | 99.35% | 446.51 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1363471MG
Encompass_Preferred
3α-Hydroxy pravastatin sodium is identified as the primary metabolite of Pravastatin. Pravastatin functions as a competitive inhibitor of HMG-CoA reductase. This product is intended solely for research purposes and is not sold for patient use.
  • Major metabolite of Pravastatin
  • Pravastatin is a competitive HMG-CoA reductase inhibitor
Catalog No. 50-003-51574