Learn More
Cayman Chemical 3'-hydroxy Puerarin, Purity: ≥98%, CAS: 117060-54-5, 25 mg
SDP
Catalog No. 502909575
Encompass_Preferred
An isoflavonoid with antioxidant activity; scavenges peroxynitrite, nitric oxide radicals, and total ROS (IC50s = 1.36, 1.13, and 6.51 µM, respectively)
Catalog No. 50-290-9575 Supplier Cayman Chemical Supplier No. 3677125 MG
An isoflavonoid with antioxidant activity; scavenges peroxynitrite, nitric oxide radicals, and total ROS (IC50s = 1.36, 1.13, and 6.51 µM, respectively)