Learn More
Medchemexpress LLC 3'-hydroxy repaglinide | 874908-14-2 | 98.1% | 468.59 | C27H36N2O5 | 5 MG
SDP
Catalog No. 5000222153
Encompass_Preferred
3'-Hydroxy repaglinide is the primary CYP2C8 metabolite of the antidiabetic drug repaglinide. It is provided as a research reference standard for metabolic, pharmacokinetic, and analytical studies, and is characterized for use in LC-MS/GC-MS assays.
  • High purity (98.09%)
  • Available in small milligram pack sizes (1 mg, 5 mg, 10 mg, 50 mg, 100 mg)
  • Suitable as a reference standard for metabolism and pharmacokinetic studies
  • Molecular formula C27H36N2O5 and molecular weight 468.59
  • Supplied and characterized for analytical method development
Catalog No. 50-002-22153 Supplier Medchemexpress LLC Supplier No. HY1353355MG
3'-Hydroxy repaglinide is the primary CYP2C8 metabolite of the antidiabetic drug repaglinide. It is provided as a research reference standard for metabolic, pharmacokinetic, and analytical studies, and is characterized for use in LC-MS/GC-MS assays.
  • High purity (98.09%)
  • Available in small milligram pack sizes (1 mg, 5 mg, 10 mg, 50 mg, 100 mg)
  • Suitable as a reference standard for metabolism and pharmacokinetic studies
  • Molecular formula C27H36N2O5 and molecular weight 468.59
  • Supplied and characterized for analytical method development