Learn More
Cayman Chemical 3-hydroxy Stearic Acid, Purity: ≥95%, CAS: 17773-30-7, 1 mg
SDP
Catalog No. 502995582
Encompass_Preferred
A long-chain saturated fatty acid; an intermediate in the production of 3-hydroxy octadecanedioic acid from stearic acid; found in C. pyloridis
Catalog No. 50-299-5582 Supplier Cayman Chemical Supplier No. 226921 MG
A long-chain saturated fatty acid; an intermediate in the production of 3-hydroxy octadecanedioic acid from stearic acid; found in C. pyloridis