Learn More
Cayman Chemical 3-hydroxy Stearic Acid, Purity: ≥95%, CAS: 17773-30-7, 25 mg
SDP
Catalog No. 502964470
Encompass_Preferred
A long-chain saturated fatty acid; an intermediate in the production of 3-hydroxy octadecanedioic acid from stearic acid; found in C. pyloridis
Catalog No. 50-296-4470 Supplier Cayman Chemical Supplier No. 2269225 MG
A long-chain saturated fatty acid; an intermediate in the production of 3-hydroxy octadecanedioic acid from stearic acid; found in C. pyloridis