Learn More
Medchemexpress LLC 3-hydroxy xylazine | 145356-33-8 | MFCD27978020 | >95.0% | 236.33 | C12H16N2OS | 5 MG
SDP
Catalog No. 5000284702
Encompass_Preferred
3-Hydroxy xylazine is a small-molecule metabolite of the α2-adrenergic receptor agonist xylazine, provided for research use in pharmacology and metabolic studies. Chemical identifiers include formula C12H16N2OS, molecular weight ≈236.33, and CAS 145356-33-8. Researchers typically use this compound for receptor binding assays, metabolic profiling, and analytical reference standards.
  • Metabolite of α2-adrenergic receptor agonist xylazine, useful in receptor pharmacology studies.
  • Known chemical identifiers for reliable database and literature matching.
  • Supplied as a solid in small research quantities suitable for in vitro and analytical work.
  • Store protected from light and under inert atmosphere; refrigerate for short-term storage.
  • Verify lot-specific certificate of analysis for purity before critical applications.
Catalog No. 50-002-84702 Supplier Medchemexpress LLC Supplier No. HYW7027125MG
3-Hydroxy xylazine is a small-molecule metabolite of the α2-adrenergic receptor agonist xylazine, provided for research use in pharmacology and metabolic studies. Chemical identifiers include formula C12H16N2OS, molecular weight ≈236.33, and CAS 145356-33-8. Researchers typically use this compound for receptor binding assays, metabolic profiling, and analytical reference standards.
  • Metabolite of α2-adrenergic receptor agonist xylazine, useful in receptor pharmacology studies.
  • Known chemical identifiers for reliable database and literature matching.
  • Supplied as a solid in small research quantities suitable for in vitro and analytical work.
  • Store protected from light and under inert atmosphere; refrigerate for short-term storage.
  • Verify lot-specific certificate of analysis for purity before critical applications.