Learn More
Medchemexpress LLC 3-hydroxyacetophenone | 121-71-1 | MFCD00002298 | >98.0% | 136.15 g/mol | C8H8O2 | 100g
SDP
Catalog No. 5000268853
Encompass_Preferred
3-Hydroxyacetophenone (m-Hydroxyacetophenone) is a hydroxy-substituted alkyl phenyl ketone and also a plant defensin in carnations 3-Hydroxyacetophenone has antifusarium activity In addition 3-Hydroxyacetophenone can be used to synthesize Rivastigmine (HY-17368)[1][2][3]
Catalog No. 50-002-68853 Supplier Medchemexpress LLC Supplier No. HYY0603100G
3-Hydroxyacetophenone (m-Hydroxyacetophenone) is a hydroxy-substituted alkyl phenyl ketone and also a plant defensin in carnations 3-Hydroxyacetophenone has antifusarium activity In addition 3-Hydroxyacetophenone can be used to synthesize Rivastigmine (HY-17368)[1][2][3]